VWR® Traceable® Bench/Portable Conductivity Meter
Supplier: VWR International
23226-505
23226-505EA 2692.54 USD
23226-505
VWR® Traceable® Bench/Portable Conductivity Meter
Electrochemistry Meters Conductivity Meters
Designed to measure conductivity in water analysis, biology, chromatography, food, electronics, dairies, and PC-board rinsing, the conductivity meter provides results in conductivity (micromhos/cm and microsiemens/cm), resistivity (megohms), and dissolved solids (parts per million).
  • Traceable®
The tough, chemical-resistant ABS housing withstands the harshest environments. The 12.7 mm (¹/₂") high 4-digit LCD readout shows temperature in °C or °F. Recorder jack allows continuous monitoring and a permanent record. The epoxy conductivity probe has a range of 1.0–200000 Micromhos.
Certifications: Meets all accreditation requirements and U.S. Government specifications. Measures reagent grade water for CAP, ASTM, NCCLS, and ACS. A serial numbered certificate is provided from an ISO 17025 calibration laboratory to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Unit complete with glass probe and 9 V alkaline battery.