Red Sable Artist Brushes, Gordon Brush
Supplier: Gordon Brush
0906-00000 0906-01000 0906-02000 0906-03000 0906-04000 0906-05000 0906-06000 0906-07000 0906-00000 0906-01000 0906-02000 0906-03000 0906-04000 0906-05000 0906-06000 0906-07000 0906-08000 0906-10000 0906-12000
22940-830EA 34.92 USD
22940-830 22940-832 22940-834 22940-836 22940-838 22940-840 22940-842 22940-844 22940-846 22940-831 22940-833 22940-835 22940-837 22940-839 22940-841 22940-843 22940-845 22940-847 22940-849 22940-851
Red Sable Artist Brushes, Gordon Brush
Brushes
Red Sable Artist Brushes are traditional round brushes made out of pure red sable hair, which is softer than camel hair, pony hair and bristle.
  • Anti-static
  • Wooden handle
These brushes are ideal for applying oils, acrylics and less caustic solutions. They are also great for applications where detail is important when working on a larger surface area.