Stainless Steel Bristle and Wood Handle Applicator Brush, Gordon Brush
Supplier: Gordon Brush
WA12SSG-12
22940-758EA 30.25 USD
22940-758
Stainless Steel Bristle and Wood Handle Applicator Brush, Gordon Brush
Brushes
A heavy-duty scrubbing duster brush that works well for getting into tight areas.
  • Known as a typewriter brush
Not recommended for use on circuit boards or SMTs as the stainless steel wire may cause unwanted scratching.