Brass Bristle and Handle Applicator Brushes, Gordon Brush
Supplier: Gordon Brush
BT201B BT202B
22940-717EA 23.02 USD
22940-717 22940-719
Brass Bristle and Handle Applicator Brushes, Gordon Brush
Brushes
These conductive applicator brushes have 0.003" soft brass wire and a brass handle.
  • Constructed with strong mechanical clamping for maximum fill retention
  • Contain no adhesives that can leach out and contaminate work areas
Conductive brushes have a surface resistivity range of 10²Ω/cm – 10⁴Ω/cm and are used to remove static buildup from the work surface. Conductive brushes will act as a conduit between the charged surface and the path to ground. Made with fully conductive materials that are active static eliminators. Use conductive brushes with a wrist strap or other mechanism to provide a path to ground.