Nylon Bristle and Zinc Plated Handle Steel Applicator Brush, Gordon Brush®
Supplier: Gordon Brush
1010NG-12
22940-714EA 119.39 USD
22940-714
Nylon Bristle and Zinc Plated Handle Steel Applicator Brush, Gordon Brush®
Brushes
Fibers are mechanically clamped into the handle for maximum fiber retention so that no adhesives can leach out and contaminate surfaces being cleaned.
  • Two sides with two end styles (tapered and flat) to fit your needs
Handle material for the 1010 series is zinc plated steel.