Tampico Bristle and Wood Shoe Handle Scratch Brush, Gordon Brush
Supplier: Gordon Brush
444T
22940-634EA 39.37 USD
22940-634
Tampico Bristle and Wood Shoe Handle Scratch Brush, Gordon Brush
Brushes
This Plater's Brush is made with fine fill material, and recommended for use on plated parts or fine metal finishing.
  • Hardwood shoe handle
Contoured for a comfortable grip.