Stainless Steel and Plywood Handle Scratch Brush, Gordon Brush
Supplier: Gordon Brush
15SSG-12
22940-624EA 43.3 USD
22940-624
Stainless Steel and Plywood Handle Scratch Brush, Gordon Brush
Brushes
The stainless steel bristle, hand-laced with stainless steel wire, remains secure even in wet environments if the wooden handle expands or contracts.
  • Toothbrush-style scratch brush
  • Hand-laced
  • Stainless steel bristle and a plywood handle
Each fill tuft is tied into position by hand, resulting in a superior quality brush with maximum fiber retention.