Stainless Steel Bristle and Plywood Scratch Brush, 1×10 Row, Gordon Brush
Supplier: Gordon Brush
11SSPG-12
22940-620EA 63.1 USD
22940-620
Stainless Steel Bristle and Plywood Scratch Brush, 1×10 Row, Gordon Brush
Brushes
Toothbrush-style scratch brush with 0.006" Stainless Steel Bristle and a Plywood Handle in a curved design for knuckle protection.
  • Brush has a 1×10 row configuration
  • Ideal for reaching narrow confned areas
  • Each fill tuft is tied into position by hand
  • Greater bristle density than staple set brushes
The stainless steel bristle, hand-laced with stainless steel wire, remains secure even in wet environments if the wooden handle expands or contracts.