Carbon Steel Bristle and Flat Wood Handle Heavy Duty Scratch Brush with Scraper, Gordon Brush®
Supplier: Gordon Brush
484CSC
22940-612EA 36.98 USD
22940-612
Carbon Steel Bristle and Flat Wood Handle Heavy Duty Scratch Brush with Scraper, Gordon Brush®
Brushes
Heavy duty brush for cleaning welds, external threads, castings and removing paint, rust and dirt.
  • Heavy duty metal scraper is included
Bristles are stapled securely into a flat hardwood handle.