SST Series Heat-Tolerant Brushes, Conductive, Gordon Brush
Supplier: Gordon Brush
SST2SSG-12 SST6SSG-12 SST12SSG-12 SST12SSG-12
22940-594EA 46.33 USD
22940-594 22940-596 22940-598 22940-712 22940-600
SST Series Heat-Tolerant Brushes, Conductive, Gordon Brush
Brushes
Stainless steel applicator brushes with a lightweight stainless steel handle.
  • Stainless steel handle is chemical resistant and non-magnetic
  • Maximum use temperature of 900°F for short term exposures
Manufactured using strong mechanical clamping for maximum fiber retention, and is adhesive-free to prevent contamination on surface areas caused by leaching.