Stainless Steel Bristle and Acetal Handle Scratch Brush, Gordon Brush
Supplier: Gordon Brush
22SSDG-12
22940-580EA 94.53 USD
22940-580
Stainless Steel Bristle and Acetal Handle Scratch Brush, Gordon Brush
Brushes
This brush has 0.003" Stainless Steel Bristle with a Delrin (Acetal) handle.
  • Hand-laced with stainless steel wire into its handle for maximum fiber retention
  • Great for removing stubborn dirt, light rust or corrosion on detailed parts
  • Cleans most tarnished metals
  • Can leave a dull finish on soft metals
Autoclavable for cleanroom, sterile, or medical applications.
Certifications: Meets FDA 21 CFR 177.2480 and ASTM D6100 standards.