Poly Hand Scraper, Perfex
Supplier: Perfex
4400
22940-046EA 10.97 USD
22940-046
Poly Hand Scraper, Perfex
Scrapers
Polypropylene scraper is ideal for countertops, walls, floors, or wherever a durable scraper is required
Won’t scratch stainless steel surfaces. No seams or cracks that might promote bacteria growth. Scraper is easy to grip, easy to clean, absorbent, and unaffected by water, acids, solvents and sanitizers. Sterilizable
Certifications: FDA and USDA approved.