PYREX® Trypsinizing Flasks with Baffles, Corning
Supplier: Corning
4460-2L
22877-184CS 573.26 USD
22877-184
PYREX® Trypsinizing Flasks with Baffles, Corning
Flasks Trypsinization Flasks
Flasks are used for converting homogenous tissue samples into cell suspension by digestion of connective proteins
Heavy beaded neck accepts cotton plugs. Four extra-deep baffles enhance vigorous agitation.