Combustion Boats, Alundum AN788, Saint Gobain
Supplier: SAINT GOBAIN ABRASIVES
3254426
22810-026EA 154.64 USD
22810-026
Combustion Boats, Alundum AN788, Saint Gobain
Boats Combustion Boats
Made of the alundum purest, 99% aluminum oxide in its basic state.
  • Temperature resistance up to 2054 °C
  • Low alkali content