Corning® Cryogenic Vial Rack, Polypropylene, 50-Position, Corning
Supplier: Corning
431131
22250-112CS 102.89 USD
22250-112
Corning® Cryogenic Vial Rack, Polypropylene, 50-Position, Corning
Racks Cryogenic Vial Racks
This reusable PP rack holds 50 cryogenic vials.
  • Locking feature for use with all Corning® self-standing vials
  • Allows one handed opening