Flux-Off® Heavy-Duty Flux Remover, ITW Chemtronics®
Supplier: ITW Chemtronics
ES1631
22221-830CS 1273.5 USD
22221-830
Flux-Off® Heavy-Duty Flux Remover, ITW Chemtronics®
Soldering Supplies Flux Removers
For the removal of all rosin and encrusted no-clean flux types from electronic assemblies, printed circuit boards, and all other electronic components
Quick-drying flux remover is nonflammable, leaves no residue, and is ozone-safe. CFC/HCFC-free. Aerosol spray.