Human Recombinant BLAME/SLAMF8 (from NS0 cells)
Supplier: R&D Systems
1907-BL-050
220023-253EA 624.74 USD
220023-253
Human Recombinant BLAME/SLAMF8 (from NS0 cells)
Proteins and Peptides
The Recombinant Human BLAME/SLAMF8 Protein from R&D Systems is derived from NS0. The Recombinant Human BLAME/SLAMF8 Protein has been validated for the following applications: Binding Activity.