Anti-TH Rabbit Polyclonal Antibody

Supplier: R&D Systems
PPS041
220022-152EA 577.02 USD
220022-152
Anti-TH Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR