Anti-GABRA4 Rabbit Polyclonal Antibody

Supplier: R&D Systems
PPS025
220022-137EA 649.32 USD
220022-137
Anti-GABRA4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen Symbol: GABRA4
Antigen Name: gamma-aminobutyric acid (GABA) A receptor, alpha 4
Antigen Synonyms: -
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Dog, Primate, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR