Anti-NR1I3 Mouse Monoclonal Antibody [clone: N4111]

Supplier: R&D Systems
PP-N4111-00
220022-108EA 678.25 USD
220022-108
Anti-NR1I3 Mouse Monoclonal Antibody [clone: N4111]
Antibodies
Type: Primary
Antigen: NR1I3
Clonality: Monoclonal
Clone: N4111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR