Anti-NR2E3 Mouse Monoclonal Antibody [clone: H7223]

Supplier: R&D Systems
PP-H7223-00
220022-079EA 692.71 USD
220022-079
Anti-NR2E3 Mouse Monoclonal Antibody [clone: H7223]
Antibodies
Direct ELISA

Type: Primary
Antigen: NR2E3
Clonality: Monoclonal
Clone: H7223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR