Anti-ESRRG Mouse Monoclonal Antibody [clone: H6812]

Supplier: R&D Systems
PP-H6812-00
220022-075EA 692.71 USD
220022-075
Anti-ESRRG Mouse Monoclonal Antibody [clone: H6812]
Antibodies
Direct ELISA

Type: Primary
Antigen: ESRRG
Clonality: Monoclonal
Clone: H6812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR