Anti-ESRRB Mouse Monoclonal Antibody [clone: H6705]

Supplier: R&D Systems
PP-H6705-00
220022-073EA 692.71 USD
220022-073
Anti-ESRRB Mouse Monoclonal Antibody [clone: H6705]
Antibodies
Type: Primary
Antigen: ESRRB
Clonality: Monoclonal
Clone: H6705
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR