Anti-THRA Mouse Monoclonal Antibody [clone: H2804]

Supplier: R&D Systems
PP-H2804-00
220022-057EA 649.32 USD
220022-057
Anti-THRA Mouse Monoclonal Antibody [clone: H2804]
Antibodies
Direct ELISA

Type: Primary
Antigen: THRA
Clonality: Monoclonal
Clone: H2804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR