Anti-IgG Goat Polyclonal Antibody (NL® 637)

Supplier: R&D Systems
NL014
220021-951EA 273.32 USD
220021-951
Anti-IgG Goat Polyclonal Antibody (NL® 637)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: NL637
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR