Anti-IgG Donkey Polyclonal Antibody (NL® 493)

Supplier: R&D Systems
NL009
220021-946EA 258.86 USD
220021-946
Anti-IgG Donkey Polyclonal Antibody (NL® 493)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: NL493
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR