Anti-AGRP Mouse Monoclonal Antibody [clone: 82815]

Supplier: R&D Systems
MAB8312
220021-590EA 822.86 USD
220021-590
Anti-AGRP Mouse Monoclonal Antibody [clone: 82815]
Antibodies
Type: Primary
Antigen: AGRP
Clonality: Monoclonal
Clone: 82815
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR