Anti-BCL2L2 Mouse Monoclonal Antibody [clone: 90316]

Supplier: R&D Systems
MAB824
220021-583EA 620.4 USD
220021-583
Anti-BCL2L2 Mouse Monoclonal Antibody [clone: 90316]
Antibodies
Type: Primary
Antigen: BCL2L2
Clonality: Monoclonal
Clone: 90316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR