Anti-CCL28 Mouse Monoclonal Antibody [clone: 62714]

Supplier: R&D Systems
MAB7171
220021-473EA 822.86 USD
220021-473
Anti-CCL28 Mouse Monoclonal Antibody [clone: 62714]
Antibodies
Type: Primary
Antigen: CCL28
Clonality: Monoclonal
Clone: 62714
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR