Anti-IL2 Mouse Monoclonal Antibody [clone: 100312]

Supplier: R&D Systems
MAB6521
220021-395EA 634.86 USD
220021-395
Anti-IL2 Mouse Monoclonal Antibody [clone: 100312]
Antibodies
Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 100312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR