Anti-NLGN3 Mouse Monoclonal Antibody [clone: 566209]

Supplier: R&D Systems
MAB6088
220021-328EA 620.4 USD
220021-328
Anti-NLGN3 Mouse Monoclonal Antibody [clone: 566209]
Antibodies
Type: Primary
Antigen Symbol: NLGN3
Antigen Name: neuroligin 3
Antigen Synonyms: HNL3
Clonality: Monoclonal
Clone: 566209
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR