Anti-AXIN2 Mouse Monoclonal Antibody [clone: 354214]

Supplier: R&D Systems
MAB6078
220021-325EA 634.86 USD
220021-325
Anti-AXIN2 Mouse Monoclonal Antibody [clone: 354214]
Antibodies
Type: Primary
Antigen: AXIN2
Clonality: Monoclonal
Clone: 354214
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR