Anti-VAMP1 Mouse Monoclonal Antibody [clone: 540524]

Supplier: R&D Systems
MAB5958
220021-299EA 620.4 USD
220021-299
Anti-VAMP1 Mouse Monoclonal Antibody [clone: 540524]
Antibodies
Type: Primary
Antigen: VAMP1
Clonality: Monoclonal
Clone: 540524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR