Anti-IL2RB Rat Monoclonal Antibody [clone: 130122]

Supplier: R&D Systems
MAB5891
220021-285EA 808.4 USD
220021-285
Anti-IL2RB Rat Monoclonal Antibody [clone: 130122]
Antibodies
Type: Primary
Antigen: IL2RB
Clonality: Monoclonal
Clone: 130122
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR