Anti-IL2 Mouse Monoclonal Antibody [clone: 144614]

Supplier: R&D Systems
MAB579
220021-256EA 837.32 USD
220021-256
Anti-IL2 Mouse Monoclonal Antibody [clone: 144614]
Antibodies
Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 144614
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Cotton Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR