Anti-HSF4 Mouse Monoclonal Antibody [clone: 504524]

Supplier: R&D Systems
MAB5695
220021-229EA 605.94 USD
220021-229
Anti-HSF4 Mouse Monoclonal Antibody [clone: 504524]
Antibodies
Type: Primary
Antigen: HSF4
Clonality: Monoclonal
Clone: 504524
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR