Anti-REG3D Rat Monoclonal Antibody [clone: 518818]

Supplier: R&D Systems
MAB5678
220021-225EA 620.4 USD
220021-225
Anti-REG3D Rat Monoclonal Antibody [clone: 518818]
Antibodies
Type: Primary
Antigen: REG3D
Clonality: Monoclonal
Clone: 518818
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR