Anti-IL5 Mouse Monoclonal Antibody [clone: 299298]

Supplier: R&D Systems
MAB5452
220021-178EA 634.86 USD
220021-178
Anti-IL5 Mouse Monoclonal Antibody [clone: 299298]
Antibodies
Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: 299298
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR