Anti-IFNL3 Mouse Monoclonal Antibody [clone: 567143]

Supplier: R&D Systems
MAB5259
220021-120EA 634.86 USD
220021-120
Anti-IFNL3 Mouse Monoclonal Antibody [clone: 567143]
Antibodies
Type: Primary
Antigen: IFNL3
Clonality: Monoclonal
Clone: 567143
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR