Anti-PPP1R2 Mouse Monoclonal Antibody [clone: 488123]

Supplier: R&D Systems
MAB4719
220020-999EA 577.02 USD
220020-999
Anti-PPP1R2 Mouse Monoclonal Antibody [clone: 488123]
Antibodies
Type: Primary
Antigen: PPP1R2
Clonality: Monoclonal
Clone: 488123
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR