Anti-PGLYRP2 Rat Monoclonal Antibody [clone: 439728]

Supplier: R&D Systems
MAB4704
220020-994EA 620.4 USD
220020-994
Anti-PGLYRP2 Rat Monoclonal Antibody [clone: 439728]
Antibodies
Type: Primary
Antigen: PGLYRP2
Clonality: Monoclonal
Clone: 439728
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR