Anti-GRM2 Mouse Monoclonal Antibody [clone: 455310]

Supplier: R&D Systems
MAB4676
220020-977EA 605.94 USD
220020-977
Anti-GRM2 Mouse Monoclonal Antibody [clone: 455310]
Antibodies
Type: Primary
Antigen Symbol: GRM2
Antigen Name: glutamate receptor, metabotropic 2
Antigen Synonyms: GLUR2, GPRC1B, MGLUR2, mGlu2
Clonality: Monoclonal
Clone: 455310
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR