Anti-RAF1 Mouse Monoclonal Antibody [clone: 563002]

Supplier: R&D Systems
MAB4540
220020-931EA 620.4 USD
220020-931
Anti-RAF1 Mouse Monoclonal Antibody [clone: 563002]
Antibodies
Type: Primary
Antigen: RAF1
Clonality: Monoclonal
Clone: 563002
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR