Anti-CDC14A Mouse Monoclonal Antibody [clone: 472508]

Supplier: R&D Systems
MAB4457
220020-901EA 620.4 USD
220020-901
Anti-CDC14A Mouse Monoclonal Antibody [clone: 472508]
Antibodies
Type: Primary
Antigen: CDC14A
Clonality: Monoclonal
Clone: 472508
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR