Anti-INSL6 Rat Monoclonal Antibody [clone: 421401]

Supplier: R&D Systems
MAB4430
220020-891EA 577.02 USD
220020-891
Anti-INSL6 Rat Monoclonal Antibody [clone: 421401]
Antibodies
Type: Primary
Antigen: INSL6
Clonality: Monoclonal
Clone: 421401
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR