Anti-ENTPD6 Mouse Monoclonal Antibody [clone: 494402]

Supplier: R&D Systems
MAB4399
220020-880EA 577.02 USD
220020-880
Anti-ENTPD6 Mouse Monoclonal Antibody [clone: 494402]
Antibodies
Type: Primary
Antigen: ENTPD6
Clonality: Monoclonal
Clone: 494402
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR