Anti-NFKBIA Mouse Monoclonal Antibody [clone: 417208]

Supplier: R&D Systems
MAB4299
220020-841EA 519.17 USD
220020-841
Anti-NFKBIA Mouse Monoclonal Antibody [clone: 417208]
Antibodies
Simple Western

Type: Primary
Antigen Symbol: NFKBIA
Antigen Name: nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha
Antigen Synonyms: IKBA, MAD-3, NFKBI
Clonality: Monoclonal
Clone: 417208
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR