Anti-JAK1 Rat Monoclonal Antibody [clone: 413104]

Supplier: R&D Systems
MAB4260
220020-823EA 663.78 USD
220020-823
Anti-JAK1 Rat Monoclonal Antibody [clone: 413104]
Antibodies
Type: Primary
Antigen: JAK1
Clonality: Monoclonal
Clone: 413104
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR