Anti-XBP1 Mouse Monoclonal Antibody [clone: 525904]

Supplier: R&D Systems
MAB4257
220020-820EA 634.86 USD
220020-820
Anti-XBP1 Mouse Monoclonal Antibody [clone: 525904]
Antibodies
Type: Primary
Antigen: XBP1
Clonality: Monoclonal
Clone: 525904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR