Anti-CFHR1 Mouse Monoclonal Antibody [clone: 442127]

Supplier: R&D Systems
MAB4247
220020-816EA 634.86 USD
220020-816
Anti-CFHR1 Mouse Monoclonal Antibody [clone: 442127]
Antibodies
Type: Primary
Antigen: CFHR1
Clonality: Monoclonal
Clone: 442127
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR